siRNA Details
| Gene Symbol: Akp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007432.2 | NP_031458.2 | 3, 4 | 306 | ||
| GenBank mRNA | BC115745.1 | - | 3, 4 | 305 | ||
| BC145627.1 | - | 2, 3 | 424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akp3| Gene Symbol: Akp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007432.2 | NP_031458.2 | 3, 4 | 306 | ||
| GenBank mRNA | BC115745.1 | - | 3, 4 | 305 | ||
| BC145627.1 | - | 2, 3 | 424 | |||