siRNA Details
| Gene Symbol: Foxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148935.2 | NP_683737.2 | 2 | 101 | ||
| GenBank mRNA | AF323488.1 | - | 2 | 109 | ||
| AY039039.1 | - | 2 | 147 | |||
| BC092242.1 | - | 2 | 90 | 30698162 | ||
| BC130222.1 | - | 2 | 102 | 40141600 | ||

Foxn4
Genomic Map