siRNA Details
| Gene Symbol: Serpinb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027548.3 | NP_081824.1 | 8 | 872 | ||
| GenBank mRNA | AF105328.1 | - | 7 | 810 | ||
| AK014524.1 | - | 8 | 875 | |||
| BC064004.1 | - | 8 | 865 | 30288309 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb7| Gene Symbol: Serpinb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027548.3 | NP_081824.1 | 8 | 872 | ||
| GenBank mRNA | AF105328.1 | - | 7 | 810 | ||
| AK014524.1 | - | 8 | 875 | |||
| BC064004.1 | - | 8 | 865 | 30288309 | ||