siRNA Details
| Gene Symbol: Amelx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081978.2 | NP_001075447.1 | 3 | 148 | ||
| NM_001290371.1 | NP_001277300.1 | 3 | 148 | |||
| NM_009666.4 | NP_033796.1 | 3 | 148 | |||
| GenBank mRNA | AK029358.1 | - | 3 | 152 | ||
| BC059090.1 | - | 3 | 118 | 30286944 | ||
| CD773419.1 | - | 3 | 152 | |||
| D31768.1 | - | 3 | 139 | |||
| D31769.1 | - | 3 | 139 | |||
| KC731591.1 | - | 3 | 148 | |||
| S74899.1 | - | 3 | 148 | |||

Amelx
Genomic Map