siRNA Details
| Gene Symbol: Ang | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161731.2 | NP_001155203.1 | 2 | 339 | ||
| NM_007447.3 | NP_031473.1 | 2 | 636 | |||
| GenBank mRNA | AK159966.1 | - | 2 | 637 | ||
| AK171251.1 | - | 2 | 267 | |||
| BC055355.1 | - | 2 | 223 | 5136641 | ||
| BF721354.1 | - | 2 | 212 | |||

Ang
Genomic Map