siRNA Details
| Gene Symbol: Asb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080444.4 | NP_536692.1 | 1 | 277 | ||
| GenBank mRNA | AF398971.1 | - | 1 | 306 | ||
| AK028538.1 | - | 1 | 277 | |||
| AK052929.1 | - | 1 | 193 | |||
| BC141311.1 | - | 1 | 268 | 9055923 | ||
| BC141313.1 | - | 1 | 268 | 9055925 | ||

Asb10
Genomic Map