siRNA Details
| Gene Symbol: Slc2a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012363.2 | NP_001012363.2 | 11 | 1229 | ||
| NM_001102414.1 | NP_001095884.1 | 11 | 1428 | |||
| NM_001102415.1 | NP_001095885.1 | 9 | 1107 | |||
| NM_145559.2 | NP_663534.1 | 10 | 1102 | |||
| GenBank mRNA | AF469480.1 | - | 11 | 1432 | ||
| AF490463.1 | - | 9 | 932 | |||
| AK033725.1 | - | 4 | 299 | |||
| AK037093.1 | - | 10 | 1093 | |||
| AK156740.1 | - | 11 | 1332 | |||
| AK172234.1 | - | 11 | 1332 | |||
| AY776155.1 | - | 11 | 1208 | |||
| BC006076.1 | - | 10 | 1102 | 3593581 | ||
| BC138213.1 | - | 12 | 1364 | 8861232 | ||
| BC145198.1 | - | 13 | 1364 | 9053749 | ||

Slc2a9
Genomic Map