siRNA Details
| Gene Symbol: Ap3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009680.3 | NP_033810.2 | 22, 23 | 2757 | ||
| GenBank mRNA | AF103809.2 | - | 22, 23 | 2694 | ||
| AK163996.1 | - | 7, 8 | 815 | |||
| BC015068.1 | - | 22, 23 | 2763 | 4018563 | ||

Ap3b1
Genomic Map