siRNA Details
| Gene Symbol: Arf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007480.1 | NP_031506.1 | 1 | 100 | ||
| GenBank mRNA | BC132452.1 | - | 2 | 88 | 40130729 | |
| BC138756.1 | - | 2 | 88 | 8861778 | ||
| D87902.1 | - | 1 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arf5| Gene Symbol: Arf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007480.1 | NP_031506.1 | 1 | 100 | ||
| GenBank mRNA | BC132452.1 | - | 2 | 88 | 40130729 | |
| BC138756.1 | - | 2 | 88 | 8861778 | ||
| D87902.1 | - | 1 | 100 | |||