siRNA Details
Gene Symbol: Phb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007531.2 | NP_031557.2 | 4 | 548 | ||
GenBank mRNA | AK011107.1 | - | 4 | 515 | ||
AK090088.1 | - | 4 | 503 | |||
AK132052.1 | - | 4 | 510 | |||
AK132058.1 | - | 4 | 537 | |||
AK135729.1 | - | 4 | 466 | |||
AK151952.1 | - | 4 | 542 | |||
AK152375.1 | - | 4 | 538 | |||
AK153368.1 | - | 4 | 538 | |||
AY211613.1 | - | 4 | 510 | |||
AY319418.1 | - | 4 | 400 | |||
BC145875.1 | - | 4 | 500 | 40131178 | ||
X78683.1 | - | 4 | 504 |
