siRNA Details
| Gene Symbol: Bmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007555.3 | NP_031581.2 | 3, 4 | 1565 | ||
| GenBank mRNA | AK033362.1 | - | 3, 4 | 1568 | ||
| AK160412.1 | - | 673 | ||||
| BC100751.1 | - | 3, 4 | 830 | |||
| BC100752.1 | - | 3, 4 | 830 | |||
| BC100753.1 | - | 3, 4 | 836 | |||
| BC100754.1 | - | 3, 4 | 830 | |||
| BC141283.1 | - | 3, 4 | 1362 | 9055895 | ||
| L41145.1 | - | 3, 4 | 831 | |||

Bmp5
Genomic Map