siRNA Details
| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009759.4 | NP_033889.2 | 15 | 1571 | ||
| GenBank mRNA | AF012104.1 | - | 14 | 1424 | ||
| AK036707.1 | - | 14 | 1430 | |||
| AK040936.1 | - | 15 | 1572 | |||
| BC138104.1 | - | 15 | 1448 | 8861123 | ||
| BC138105.1 | - | 15 | 1448 | 8861124 | ||
| U88091.1 | - | 15 | 1557 | |||

Bmx
Genomic Map