siRNA Details
| Gene Symbol: Smyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160127.1 | NP_001153599.1 | 8, 9 | 1225 | ||
| NM_009762.2 | NP_033892.2 | 7, 8 | 1186 | |||
| GenBank mRNA | AK076266.1 | - | 7, 8 | 1151 | ||
| AK142153.1 | - | 7, 8 | 1154 | |||
| AK142252.1 | - | 8, 9 | 1203 | |||
| BC076601.1 | - | 8, 9 | 1203 | 30609878 | ||
| U76371.1 | - | 1330 | ||||
| U76373.2 | - | 8, 9 | 1197 | |||
| U76374.2 | - | 7, 8 | 1158 | |||

Smyd1
Genomic Map