siRNA Details
| Gene Symbol: Zfp36l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007564.5 | NP_031590.1 | 2 | 663 | ||
| GenBank mRNA | AK036950.1 | - | 2 | 664 | ||
| AK051035.1 | - | 2 | 664 | |||
| BC016621.1 | - | 2 | 648 | 3157001 | ||
| M58566.1 | - | 2 | 592 | |||

Zfp36l1
Genomic Map