siRNA Details
| Gene Symbol: Btk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013482.2 | NP_038510.2 | 12 | 1228 | ||
| GenBank mRNA | BC053392.1 | - | 12 | 1164 | 30075322 | |
| L08967.1 | - | 12 | 1204 | |||
| L10627.1 | - | 12 | 1239 | |||
| L29788.1 | - | 12 | 1200 | |||

Btk
Genomic Map