siRNA Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301250.1 | NP_001288179.1 | 12, 13 | 1832 | ||
| NM_007602.4 | NP_031628.1 | 12, 13 | 1996 | |||
| GenBank mRNA | AK139476.1 | - | 1, 2 | 145 | ||
| AK155179.1 | - | 12, 13 | 1974 | |||
| AK164293.1 | - | 12, 13 | 1906 | |||
| AK170034.1 | - | 12, 13 | 1933 | |||
| BC014767.1 | - | 12, 13 | 1824 | 4458488 | ||
| U85020.1 | - | 12, 13 | 1915 | |||
| Y10656.1 | - | 12, 13 | 1928 | |||

Capn5
Genomic Map