siRNA Details
| Gene Symbol: Cbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007622.3 | NP_031648.1 | 3 | 431 | ||
| GenBank mRNA | AK014215.1 | - | 3 | 407 | ||
| AK017608.1 | - | 3 | 320 | |||
| AK157710.1 | - | 3 | 433 | |||
| BC055290.1 | - | 3 | 374 | 6313666 | ||
| X56690.1 | - | 2 | 176 | |||

Cbx1
Genomic Map