siRNA Details
| Gene Symbol: Cbx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076789.1 | NP_001070257.1 | 4 | 535 | ||
| NM_001110216.1 | NP_001103686.1 | 5 | 636 | |||
| NM_007626.3 | NP_031652.1 | 4 | 536 | |||
| GenBank mRNA | AK008792.1 | - | 4 | 571 | ||
| AK018349.1 | - | 4 | 482 | |||
| AK030366.1 | - | 4 | 536 | |||
| AK030442.1 | - | 5 | 589 | |||
| AK032975.1 | - | 4 | 492 | |||
| BC004707.1 | - | 4 | 535 | 3499489 | ||
| X99641.1 | - | 4 | 468 | |||

Cbx5
Genomic Map