siRNA Details
Gene Symbol: Cd34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111059.1 | NP_001104529.1 | 1, 2 | 217 | ||
NM_133654.3 | NP_598415.1 | 1, 2 | 217 | |||
GenBank mRNA | AK044121.1 | - | 1, 2 | 153 | ||
AK051507.1 | - | 1, 2 | 133 | |||
AK053874.1 | - | 1, 2 | 153 | |||
AK089010.1 | - | 1, 2 | 123 | |||
AK167329.1 | - | 1, 2 | 128 | |||
AK167350.1 | - | 1, 2 | 128 | |||
BC006607.1 | - | 1, 2 | 133 | 3966337 | ||
BC066820.1 | - | 1, 2 | 90 | |||
BY249301.1 | - | 1, 2 | 220 | |||
BY327943.1 | - | 1, 2 | 170 | |||
S69293.1 | - | 1, 2 | 95 | |||
S69299.1 | - | 1, 2 | 72 | |||
S69301.1 | - | 1, 2 | 72 |
