siRNA Details
| Gene Symbol: Cd34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111059.1 | NP_001104529.1 | 3 | 497 | ||
| NM_133654.3 | NP_598415.1 | 3 | 497 | |||
| GenBank mRNA | AK044121.1 | - | 3 | 433 | ||
| AK051507.1 | - | 2 | 1711 | |||
| AK053874.1 | - | 3 | 433 | |||
| AK089010.1 | - | 3 | 403 | |||
| AK167329.1 | - | 3 | 408 | |||
| AK167350.1 | - | 3 | 408 | |||
| BC006607.1 | - | 3 | 413 | 3966337 | ||
| S69293.1 | - | 3 | 375 | |||
| S69299.1 | - | 3 | 352 | |||
| S69301.1 | - | 3 | 352 | |||

Cd34
Genomic Map