siRNA Details
| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037366.1 | - | 10 | 1550 | ||
| AK028625.1 | - | 7 | 1216 | |||
| AK029512.1 | - | 10 | 1596 | |||
| AK030601.1 | - | 10 | 1576 | |||
| BC011278.1 | - | 10 | 1561 | 3674983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entpd1| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037366.1 | - | 10 | 1550 | ||
| AK028625.1 | - | 7 | 1216 | |||
| AK029512.1 | - | 10 | 1596 | |||
| AK030601.1 | - | 10 | 1576 | |||
| BC011278.1 | - | 10 | 1561 | 3674983 | ||