siRNA Details
| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037366.1 | - | 9 | 1367 | ||
| AK029512.1 | - | 9 | 1413 | |||
| AK030601.1 | - | 9 | 1393 | |||
| BC011278.1 | - | 9 | 1378 | 3674983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entpd1| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037366.1 | - | 9 | 1367 | ||
| AK029512.1 | - | 9 | 1413 | |||
| AK030601.1 | - | 9 | 1393 | |||
| BC011278.1 | - | 9 | 1378 | 3674983 | ||