siRNA Details
| Gene Symbol: Cd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007650.3 | NP_031676.3 | 5 | 652 | ||
| GenBank mRNA | AK042105.1 | - | 5 | 596 | ||
| AK089076.1 | - | 5 | 601 | |||
| AK143736.1 | - | 5 | 628 | |||
| BC011258.1 | - | 5 | 652 | 4188683 | ||
| M15177.1 | - | 5 | 530 | |||
| U65961.1 | - | 5 | 530 | |||

Cd5
Genomic Map