siRNA Details
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007651.3 | NP_031677.1 | 2 | 211 | ||
GenBank mRNA | AA177409.1 | - | 2 | 99 | ||
AK156510.1 | - | 2 | 118 | |||
AK160326.1 | - | 2 | 109 | |||
BC021310.1 | - | 2 | 125 | 5010267 | ||
BC052905.1 | - | 2 | 76 | 30078163 | ||
X97227.1 | - | 1 | 12 |
