siRNA Details
| Gene Symbol: Cd53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007651.3 | NP_031677.1 | 8 | 816 | ||
| GenBank mRNA | AK156510.1 | - | 8 | 723 | ||
| AK160326.1 | - | 8 | 714 | |||
| BC021310.1 | - | 8 | 730 | 5010267 | ||
| BC052905.1 | - | 8 | 681 | 30078163 | ||
| X97227.1 | - | 7 | 617 | |||

Cd53
Genomic Map