siRNA Details
| Gene Symbol: Cdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009870.3 | NP_034000.1 | 4 | 547 | ||
| GenBank mRNA | AK003804.1 | - | 4 | 514 | ||
| AK011871.1 | - | 4 | 535 | |||
| AK012064.1 | - | 4 | 516 | |||
| AK078440.1 | - | 4 | 501 | |||
| AK146107.1 | - | 4 | 514 | |||
| AK153465.1 | - | 4 | 547 | |||
| BC031599.1 | - | 4 | 536 | |||
| BC046336.1 | - | 4 | 547 | 6312184 | ||
| BC052694.1 | - | 4 | 542 | 5714971 | ||
| L01640.1 | - | 3 | 374 | |||

Cdk4
Genomic Map