siRNA Details
| Gene Symbol: Cdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009870.3 | NP_034000.1 | 5, 6 | 813 | ||
| GenBank mRNA | AK003804.1 | - | 5, 6 | 780 | ||
| AK011871.1 | - | 5, 6 | 801 | |||
| AK012064.1 | - | 5, 6 | 782 | |||
| AK019380.1 | - | 5, 6 | 766 | |||
| AK078440.1 | - | 5, 6 | 767 | |||
| AK146107.1 | - | 5, 6 | 780 | |||
| AK153465.1 | - | 5, 6 | 813 | |||
| AK218927.1 | - | 1, 2 | 100 | |||
| BC046336.1 | - | 5, 6 | 813 | 6312184 | ||
| BC052694.1 | - | 5, 6 | 808 | 5714971 | ||
| L01640.1 | - | 4, 5 | 640 | |||

Cdk4
Genomic Map