siRNA Details
| Gene Symbol: Cdr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007672.2 | NP_031698.2 | 3 | 473 | ||
| GenBank mRNA | AK148057.1 | - | 3 | 457 | ||
| AK171706.1 | - | 3 | 428 | |||
| BC117530.1 | - | 3 | 362 | 40106297 | ||
| BC118511.1 | - | 3 | 362 | 40106298 | ||
| BY713743.1 | - | 3 | 473 | |||
| U88588.1 | - | 4 | 366 | |||

Cdr2
Genomic Map