siRNA Details
| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009914.4 | NP_034044.3 | 2 | 577 | ||
| GenBank mRNA | AK041106.1 | - | 2 | 578 | ||
| AK089875.1 | - | 2 | 578 | |||
| AK089895.1 | - | 2 | 449 | |||
| AK153518.1 | - | 2 | 578 | |||
| BC108967.2 | - | 2 | 403 | 40047217 | ||
| BC108968.1 | - | 2 | 403 | 40047218 | ||
More Information
Other Species Targeted
Set Membership: |
|

Ccr3
Genomic Map