siRNA Details
| Gene Symbol: Col2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113515.2 | NP_001106987.2 | 52 | 4193 | ||
| NM_031163.3 | NP_112440.2 | 53 | 4397 | |||
| GenBank mRNA | AK028295.1 | - | 27 | 2379 | ||
| BC030913.1 | - | 23 | 2185 | |||
| BC051383.1 | - | 52 | 4104 | 6312433 | ||
| BC052326.1 | - | 52 | 4083 | 6315379 | ||
| BC082331.1 | - | 53 | 4330 | 30537724 | ||

Col2a1
Genomic Map