siRNA Details
| Gene Symbol: Col2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113515.2 | NP_001106987.2 | 51 | 4043 | ||
| NM_031163.3 | NP_112440.2 | 52 | 4247 | |||
| GenBank mRNA | AK028295.1 | - | 26 | 2229 | ||
| BC030913.1 | - | 22 | 2035 | |||
| BC051383.1 | - | 51 | 3954 | 6312433 | ||
| BC052326.1 | - | 51 | 3933 | 6315379 | ||
| BC082331.1 | - | 52 | 4180 | 30537724 | ||

Col2a1
Genomic Map