siRNA Details
| Gene Symbol: Col2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113515.2 | NP_001106987.2 | 51, 52 | 4107 | ||
| NM_031163.3 | NP_112440.2 | 52, 53 | 4311 | |||
| GenBank mRNA | AK028295.1 | - | 26, 27 | 2293 | ||
| BC030913.1 | - | 22, 23 | 2099 | |||
| BC051383.1 | - | 51, 52 | 4018 | 6312433 | ||
| BC052326.1 | - | 51, 52 | 3997 | 6315379 | ||
| BC082331.1 | - | 52, 53 | 4244 | 30537724 | ||

Col2a1
Genomic Map