siRNA Details
| Gene Symbol: Col4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009932.3 | NP_034062.3 | 14, 15 | 1185 | ||
| GenBank mRNA | AK053858.1 | - | 14, 15 | 1185 | ||
| BC138040.1 | - | 13, 14 | 897 | 8861059 | ||
| J04695.1 | - | 14, 15 | 990 | |||

Col4a2
Genomic Map