siRNA Details
| Gene Symbol: Col4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009932.3 | NP_034062.3 | 25, 26 | 2300 | ||
| GenBank mRNA | BC138040.1 | - | 24, 25 | 2012 | 8861059 | |
| J04695.1 | - | 25, 26 | 2105 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col4a2| Gene Symbol: Col4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009932.3 | NP_034062.3 | 25, 26 | 2300 | ||
| GenBank mRNA | BC138040.1 | - | 24, 25 | 2012 | 8861059 | |
| J04695.1 | - | 25, 26 | 2105 | |||