siRNA Details
| Gene Symbol: Atf6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017406.4 | NP_059102.2 | 4 | 387 | ||
| GenBank mRNA | AK090378.1 | - | 4 | 334 | ||
| AK143765.1 | - | 4 | 320 | |||
| AK146671.1 | - | 4 | 354 | |||
| AK159169.1 | - | 4 | 320 | |||
| AK170027.1 | - | 4 | 320 | |||
| BC013534.1 | - | 4 | 387 | 4239106 | ||
| BC052635.1 | - | 4 | 322 | 6395626 | ||

Atf6b
Genomic Map