siRNA Details
| Gene Symbol: Crkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007764.5 | NP_031790.2 | 2 | 1065 | ||
| GenBank mRNA | AK048641.1 | - | 2 | 1063 | ||
| AK052315.1 | - | 2 | 1065 | |||
| AK163981.1 | - | 2 | 1037 | |||
| BC131984.1 | - | 2 | 832 | 40130261 | ||
| BC131986.1 | - | 2 | 832 | 40130263 | ||
| X90648.1 | - | 2 | 904 | |||

Crkl
Genomic Map