siRNA Details
Gene Symbol: Crybb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023695.2 | NP_076184.1 | 5 | 560 | ||
GenBank mRNA | AF106853.1 | - | 5 | 560 | ||
AK014012.1 | - | 5 | 562 | |||
AK053869.1 | - | 5 | 561 | |||
BC029012.1 | - | 4 | 544 | 5358714 |
