siRNA Details
Gene Symbol: Crybb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159650.1 | NP_001153122.1 | 5 | 442 | ||
NM_021352.3 | NP_067327.1 | 5 | 512 | |||
GenBank mRNA | AJ272229.2 | - | 4 | 496 | ||
AK053552.1 | - | 3 | 826 | |||
BC031442.1 | - | 4 | 503 | 4501690 | ||
BC058522.1 | - | 4 | 421 | 6824229 | ||
CJ046471.1 | - | 5 | 513 |
