siRNA Details
| Gene Symbol: Crybb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159650.1 | NP_001153122.1 | 4 | 361 | ||
| NM_021352.3 | NP_067327.1 | 4 | 431 | |||
| GenBank mRNA | AJ272229.2 | - | 3 | 415 | ||
| AK053552.1 | - | 2 | 745 | |||
| BC031442.1 | - | 3 | 422 | 4501690 | ||
| BC058522.1 | - | 3 | 340 | 6824229 | ||
| CJ046471.1 | - | 4 | 432 | |||

Crybb3
Genomic Map