siRNA Details
| Gene Symbol: Cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026444.3 | NP_080720.1 | 8 | 895 | ||
| GenBank mRNA | AB056479.1 | - | 8 | 809 | ||
| AK010298.1 | - | 8 | 982 | |||
| AK012104.1 | - | 2 | 73 | |||
| AK012151.1 | - | 8 | 926 | |||
| AK145643.1 | - | 8 | 927 | |||
| AK149990.1 | - | 8 | 994 | |||
| AK163273.1 | - | 8 | 920 | |||
| AK166814.1 | - | 8 | 930 | |||
| AK167125.1 | - | 8 | 927 | |||
| BC013554.1 | - | 8 | 910 | 4237940 | ||
| BC029754.1 | - | 8 | 895 | 2655044 | ||
| DQ403125.1 | - | 2 | 143 | |||

Cs
Genomic Map