siRNA Details
| Gene Symbol: Pcyt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163159.1 | NP_001156631.1 | 5, 6 | 765 | ||
| NM_001163160.1 | NP_001156632.1 | 5, 6 | 699 | |||
| NM_009981.4 | NP_034111.1 | 5, 6 | 601 | |||
| GenBank mRNA | AK044585.1 | - | 5, 6 | 602 | ||
| AK076050.1 | - | 5, 6 | 526 | |||
| AK076830.1 | - | 5, 6 | 700 | |||
| BC018313.1 | - | 4, 5 | 484 | 4162053 | ||
| L28956.1 | - | 5, 6 | 570 | |||
| Z12302.1 | - | 5, 6 | 560 | |||

Pcyt1a
Genomic Map