siRNA Details
| Gene Symbol: Cyp2c29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007815.3 | NP_031841.3 | 9 | 1375 | ||
| GenBank mRNA | AK139760.1 | - | 1415 | |||
| AK149558.1 | - | 9 | 1194 | |||
| BC019908.1 | - | 9 | 1357 | 5123631 | ||
| D17674.1 | - | 9 | 1349 | |||

Cyp2c29
Genomic Map