siRNA Details
| Gene Symbol: Cyp2d10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010005.3 | NP_034135.2 | 6 | 975 | ||
| GenBank mRNA | BC010989.1 | - | 6 | 973 | 4193743 | |
| BC051039.1 | - | 6 | 952 | |||
| BC057924.1 | - | 6 | 962 | 5100459 | ||
| BF660186.1 | - | 4 | 411 | |||
| CB951489.1 | - | 2 | 204 | |||
| M27167.1 | - | 6 | 953 | |||

Cyp2d10
Genomic Map