siRNA Details
| Gene Symbol: Cyp2d9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010006.2 | NP_034136.2 | 6 | 963 | ||
| GenBank mRNA | AK143963.1 | - | 6 | 963 | ||
| BC010593.1 | - | 6 | 938 | 3964178 | ||
| BC094015.1 | - | 6 | 946 | 5050233 | ||
| M23998.1 | - | 6 | 959 | |||
| M27168.1 | - | 6 | 938 | |||

Cyp2d9
Genomic Map