siRNA Details
| Gene Symbol: Ddx3x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010028.3 | NP_034158.1 | 13 | 1460 | ||
| GenBank mRNA | AK140990.1 | - | 13 | 1461 | ||
| AK163250.1 | - | 13 | 1435 | |||
| AK176969.1 | - | 3 | 243 | |||
| AK205639.1 | - | 3 | 244 | |||
| BC150862.1 | - | 13 | 1414 | 9087773 | ||
| BC172016.1 | - | 13 | 1411 | 9054302 | ||
| DQ858457.1 | - | 3 | 249 | |||
| DQ858458.1 | - | 3 | 249 | |||
| L25126.1 | - | 13 | 1392 | |||
| Z38117.1 | - | 13 | 1444 | |||

Ddx3x
Genomic Map