siRNA Details
| Gene Symbol: Dhcr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007856.2 | NP_031882.1 | 4 | 614 | ||
| GenBank mRNA | AF057368.1 | - | 3 | 146 | ||
| AK146811.1 | - | 3 | 137 | |||
| BC006854.1 | - | 2 | 128 | 3157987 | ||
| CT010277.1 | - | 2 | 107 | |||

Dhcr7
Genomic Map