siRNA Details
| Gene Symbol: Dmpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190490.1 | NP_001177419.1 | 4 | 506 | ||
| NM_001190491.1 | NP_001177420.1 | 4 | 506 | |||
| NM_032418.2 | NP_115794.1 | 4 | 506 | |||
| GenBank mRNA | BC024150.1 | - | 4 | 493 | 4217643 | |
| BC056615.1 | - | 4 | 485 | |||
| BC075715.1 | - | 4 | 414 | 4989259 | ||
| S60313.1 | - | 1 | 13 | |||
| S60314.1 | - | 1 | 13 | |||
| S60315.1 | - | 1 | 13 | |||
| S60316.1 | - | 1 | 13 | |||

Dmpk
Genomic Map