siRNA Details
Gene Symbol: F630201L12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155102.1 | - | 1 | 1650 |
Gene Symbol: Dok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010071.2 | NP_034201.1 | 2 | 154 | ||
GenBank mRNA | AF030627.1 | - | 2 | 154 | ||
AF035117.1 | - | 2 | 150 | |||
AF059583.1 | - | 2 | 140 | |||
AK155579.1 | - | 2 | 151 | |||
AK158246.1 | - | 2 | 118 | |||
AK159466.1 | - | 2 | 195 | |||
AK159472.1 | - | 2 | 232 | |||
BC004590.1 | - | 2 | 146 | 3587682 |
