siRNA Details
| Gene Symbol: Prl8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289919.1 | NP_001276848.1 | 3 | 288 | ||
| NM_010088.2 | NP_034218.1 | 3 | 291 | |||
| GenBank mRNA | AF011385.1 | - | 3 | 275 | ||
| AF015729.1 | - | 3 | 285 | |||
| AK131668.1 | - | 3 | 289 | |||
| AK146049.1 | - | 3 | 292 | |||
| BC061150.1 | - | 3 | 288 | 30317747 | ||

Prl8a2
Genomic Map