siRNA Details
Gene Symbol: S1pr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010101.4 | NP_034231.1 | 2 | 1209 | ||
GenBank mRNA | AB028143.1 | - | 2 | 1077 | ||
AK028043.1 | - | 2 | 1066 | |||
AK029852.1 | - | 2 | 1065 | |||
AK030134.1 | - | 2 | 1064 | |||
AK047268.1 | - | 2 | 1057 | |||
AK078443.1 | - | 2 | 1063 | |||
AK081919.1 | - | 2 | 1054 | |||
AK083405.1 | - | 2 | 1043 | |||
AK084944.1 | - | 2 | 1064 | |||
AK085180.1 | - | 2 | 1064 | |||
AK087076.1 | - | 2 | 1064 | |||
AK087139.1 | - | 2 | 1063 | |||
AK209260.1 | - | 1 | 58 | |||
BC064007.1 | - | 2 | 1049 | 30244496 | ||
BC068176.1 | - | 2 | 1049 | 30613259 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
